Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL13RA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001560.2 | NP_001551.1 | 7 | 888 | ||
XM_011531336.1 | XP_011529638.1 | 7 | 1166 | |||
XM_017029507.1 | XP_016884996.1 | 6 | 905 | |||
GenBank mRNA | AB209849.1 | - | 7 | 843 | ||
AK296958.1 | - | 2 | 117 | |||
AK313467.1 | - | 7 | 877 | |||
BC009960.2 | - | 7 | 875 | 4300487 | ||
BC015768.1 | - | 7 | 892 | 4868206 | ||
U81379.3 | - | 7 | 885 | |||
Y09328.1 | - | 7 | 878 | |||
Y10659.1 | - | 7 | 888 |