Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St6gal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252505.1 | NP_001239434.1 | 8 | 1624 | ||
NM_001252506.1 | NP_001239435.1 | 7 | 1491 | |||
NM_145933.4 | NP_666045.1 | 8 | 1955 | |||
GenBank mRNA | AK034768.1 | - | 2 | 301 | ||
AK084124.1 | - | 8 | 1790 | |||
BC027833.1 | - | 5 | 1152 | 5321763 | ||
BC092222.1 | - | 8 | 1933 | 6844069 | ||
BC096026.1 | - | 8 | 1603 | 6833540 | ||
D16106.1 | - | 7 | 1442 |