Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St3gal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009177.4 | NP_033203.1 | 8, 9 | 1829 | ||
GenBank mRNA | AK031344.1 | - | 8, 9 | 1719 | ||
AK078469.1 | - | 7, 8 | 1138 | |||
AK138747.1 | - | 8, 9 | 1778 | |||
BC023130.1 | - | 7, 8 | 2081 | |||
BC040763.1 | - | 5, 6 | 882 | |||
BC084730.1 | - | 5, 6 | 1030 | 30355523 | ||
BC099670.1 | - | 8, 9 | 1716 | 4187038 | ||
BC099693.1 | - | 9, 10 | 1796 | 4910140 | ||
X73523.1 | - | 5, 6 | 847 |