Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009181.2 | NP_033207.2 | 1 | 267 | ||
GenBank mRNA | BB649753.1 | - | 1 | 267 | ||
BC096546.1 | - | 1 | 259 | 6833487 | ||
X83562.1 | - | 1 | 246 |