Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009182.3 | NP_033208.2 | 5 | 1148 | ||
GenBank mRNA | AK015874.1 | - | 5 | 1132 | ||
AK133780.1 | - | 3 | 433 | |||
AK134678.1 | - | 3 | 444 | |||
AK161482.1 | - | 5 | 1149 | |||
BC075645.1 | - | 3 | 443 | 30620472 | ||
X80502.1 | - | 3 | 433 |