Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tra2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009186.4 | NP_033212.1 | 4 | 640 | ||
GenBank mRNA | AK049573.1 | - | 4 | 641 | ||
AK152519.1 | - | 4 | 635 | |||
AK152761.1 | - | 4 | 641 | |||
AK167387.1 | - | 4 | 639 | |||
AK167544.1 | - | 4 | 641 | |||
AK182183.1 | - | 1 | 19 | |||
BC057996.1 | - | 5 | 816 | |||
BC061177.1 | - | 4 | 637 | 6594226 | ||
BC086795.1 | - | 4 | 627 | 6828358 | ||
X80232.1 | - | 4 | 632 |