Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000585.4 | NP_000576.1 | 7 | 661 | ||
NM_172175.2 | NP_751915.1 | 9 | 982 | |||
NR_037840.2 | 7 | 1137 | ||||
GenBank mRNA | AF031167.1 | - | 6 | 724 | ||
AK122993.1 | - | 6 | 5355 | |||
AK290619.1 | - | 11 | 954 | |||
AY720442.1 | - | 4 | 217 | |||
BC018149.2 | - | 7 | 1131 | 3851514 | ||
BC100961.1 | - | 6 | 486 | 40004735 | ||
BC100962.1 | - | 6 | 486 | 40004736 | ||
BC100963.1 | - | 5 | 367 | 40004738 | ||
CR541980.1 | - | 4 | 206 | |||
CR542007.1 | - | 4 | 206 | |||
U14407.1 | - | 7 | 603 | |||
X94222.1 | - | 6 | 428 | |||
X94223.1 | - | 6 | 428 | |||
Z38000.1 | - | 4 | 253 |