Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IL15RA SIRNA Assays ›
Gene Symbol:
IL15RA
Gene Name:
interleukin 15 receptor subunit alpha
Gene Aliases:
CD215
Chromosome Location:
Chr.10: 5948897 - 5978741 on Build GRCh38
UniGene ID:
Hs.445124
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s7380
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IL15RA
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001243539.1
NP_001230468.1
2
496
NM_001256765.1
NP_001243694.1
3
445
NM_002189.3
NP_002180.1
2
292
NM_172200.2
NP_751950.2
2
292
XM_011519461.2
XP_011517763.1
4
686
XM_011519462.2
XP_011517764.1
3
604
XM_011519463.2
XP_011517765.1
4
686
XM_011519464.2
XP_011517766.1
3
533
XM_011519465.1
XP_011517767.1
4
686
XM_011519466.2
XP_011517768.1
4
686
XM_011519467.2
XP_011517769.1
2
449
XM_011519468.1
XP_011517770.1
4
686
XM_011519469.1
XP_011517771.1
4
686
XM_011519470.2
XP_011517772.1
2
451
XM_017016195.1
XP_016871684.1
3
533
XM_017016196.1
XP_016871685.1
2
258
XM_017016197.1
XP_016871686.1
3
273
XM_017016199.1
XP_016871688.1
4
686
XM_017016200.1
XP_016871689.1
2
943
XM_017016201.1
XP_016871690.1
3
640
XM_017016202.1
XP_016871691.1
3
262
XM_017016203.1
XP_016871692.1
2
943
GenBank mRNA
AF035279.1
-
2
227
AK304211.1
-
2
203
BC074726.2
-
2
204
30915179
BC107777.1
-
2
360
6497994
BC121140.2
-
2
199
40120998
BC121141.2
-
2
204
40120999
CR457064.1
-
1
65
CR542023.1
-
2
173
DC298692.1
-
2
297
HQ401283.1
-
3
326
JX987304.1
-
3
326
JX987305.1
-
3
326
JX987306.1
-
3
326
JX987307.1
-
3
326
JX987308.1
-
3
326
JX987309.1
-
3
326
JX987310.1
-
3
326
JX987311.1
-
3
326
U31628.1
-
2
255
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
cell proliferation
cytokine-mediated signaling pathway
signal transducer activity
cytokine receptor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-zpscg:80/10.239.160.225:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0