Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009190.2 | NP_033216.2 | 8, 9 | 1154 | ||
GenBank mRNA | AK151023.1 | - | 8, 9 | 1108 | ||
AK152131.1 | - | 8, 9 | 1141 | |||
AK165594.1 | - | 1099 | ||||
AK168270.1 | - | 8, 9 | 1045 | |||
BC003799.1 | - | 8, 9 | 1061 | 3488471 | ||
U10119.1 | - | 8, 9 | 1105 |