Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011390.2 | NP_035520.1 | 22 | 2930 | ||
GenBank mRNA | AF087436.1 | - | 22 | 2932 | ||
AK131129.1 | - | 22 | 2884 | |||
AK143535.1 | - | 22 | 2924 | |||
AK166215.1 | - | 22 | 2924 | |||
AK169950.1 | - | 22 | 2920 | |||
BC059242.1 | - | 22 | 2922 | |||
BC060133.1 | - | 22 | 2922 | |||
BC079677.1 | - | 18 | 4787 | |||
BC141107.1 | - | 22 | 2890 | 9055719 |