Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009197.2 | NP_033223.2 | 4 | 1355 | ||
GenBank mRNA | AB265788.1 | - | 4 | 1181 | ||
AF045692.1 | - | 4 | 1403 | |||
BC080678.1 | - | 4 | 1191 | 6313495 | ||
BC096587.1 | - | 4 | 1291 | 3992886 |