Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc34a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011392.2 | NP_035522.2 | 3 | 321 | ||
GenBank mRNA | AB163921.1 | - | 326 | |||
AB163922.1 | - | 3 | 265 | |||
AK018740.2 | - | 3 | 276 | |||
BC020529.1 | - | 3 | 264 | 4223484 | ||
CB952922.1 | - | 3 | 324 | |||
L33878.1 | - | 2 | 248 | |||
U22465.1 | - | 3 | 265 |