Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc19a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199271.1 | NP_001186200.1 | 5 | 1303 | ||
NM_031196.3 | NP_112473.1 | 5 | 1397 | |||
GenBank mRNA | AK002560.1 | - | 5 | 1314 | ||
AK050714.1 | - | 5 | 1427 | |||
AK079592.1 | - | 5 | 1358 | |||
AK085024.2 | - | 5 | 1345 | |||
AK088803.1 | - | 5 | 1360 | |||
AK138577.1 | - | 5 | 1223 | |||
BC015263.1 | - | 5 | 1315 | 4239690 | ||
L23755.1 | - | 4 | 1255 | |||
L36539.1 | - | 5 | 1310 | |||
U32469.1 | - | 5 | 1337 | |||
U66103.1 | - | 5 | 1350 |