Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc19a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199271.1 | NP_001186200.1 | 3 | 804 | ||
NM_031196.3 | NP_112473.1 | 3 | 898 | |||
GenBank mRNA | AK002560.1 | - | 3 | 815 | ||
AK050714.1 | - | 3 | 928 | |||
AK079592.1 | - | 3 | 859 | |||
AK085024.2 | - | 3 | 845 | |||
AK088803.1 | - | 3 | 861 | |||
AK182397.1 | - | 1 | 19 | |||
AK182822.1 | - | 1 | 19 | |||
BC015263.1 | - | 3 | 816 | 4239690 | ||
L23755.1 | - | 2 | 756 | |||
L36539.1 | - | 3 | 811 | |||
U32469.1 | - | 3 | 838 | |||
U66103.1 | - | 3 | 851 |