Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077514.3 | NP_001070982.1 | 8 | 1785 | ||
NM_001077515.2 | NP_001070983.1 | 8 | 1212 | |||
NM_011393.2 | NP_035523.1 | 8 | 1212 | |||
GenBank mRNA | AB007810.1 | - | 8 | 1842 | ||
AB007811.1 | - | 8 | 1201 | |||
AB007812.1 | - | 8 | 1212 | |||
AK134609.1 | - | 8 | 1786 | |||
AK139042.1 | - | 2 | 273 | |||
AK140095.1 | - | 8 | 1780 | |||
BC138255.1 | - | 8 | 1212 | 8861274 | ||
BC138256.1 | - | 8 | 1212 | 8861275 | ||
D43796.1 | - | 8 | 1365 | |||
KC825358.1 | - | 7 | 1174 | |||
U11763.1 | - | 8 | 1370 | |||
U24699.1 | - | 7 | 1183 | |||
U75372.1 | - | 8 | 1798 |