Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148938.3 | NP_683740.1 | 8 | 1844 | ||
GenBank mRNA | AF330257.1 | - | 7 | 1344 | ||
AK031831.1 | - | 8 | 1507 | |||
AK040798.1 | - | 8 | 1504 | |||
AK045949.1 | - | 8 | 1523 | |||
AK049090.1 | - | 8 | 1504 | |||
AK080879.1 | - | 8 | 1528 | |||
AK082736.1 | - | 8 | 1504 | |||
AK084968.1 | - | 8 | 1488 | |||
AK158777.1 | - | 7 | 1378 | |||
BC055300.1 | - | 4 | 2675 | |||
BC058711.1 | - | 8 | 1767 | 6842500 | ||
BC066154.1 | - | 8 | 1447 | 30102325 |