Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148938.3 | NP_683740.1 | 4 | 941 | ||
GenBank mRNA | AF330257.1 | - | 3 | 441 | ||
AK031831.1 | - | 4 | 601 | |||
AK040798.1 | - | 4 | 601 | |||
AK045949.1 | - | 4 | 620 | |||
AK049090.1 | - | 4 | 601 | |||
AK080879.1 | - | 4 | 625 | |||
AK082736.1 | - | 4 | 601 | |||
AK084968.1 | - | 4 | 585 | |||
AK158777.1 | - | 3 | 475 | |||
BC055300.1 | - | 1 | 509 | |||
BC058711.1 | - | 4 | 864 | 6842500 | ||
BC066154.1 | - | 4 | 544 | 30102325 |