Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc1a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009200.3 | NP_033226.1 | 9 | 1656 | ||
GenBank mRNA | AK159197.1 | - | 9 | 1657 | ||
BB668161.1 | - | 1 | 39 | |||
BC138120.1 | - | 9 | 1558 | 8861139 | ||
BC138140.1 | - | 9 | 1558 | 8861159 | ||
D83262.1 | - | 9 | 1650 |