Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc20a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011394.3 | NP_035524.2 | 4 | 974 | ||
GenBank mRNA | AF196476.1 | - | 3 | 742 | ||
AK076380.1 | - | 1 | 1634 | |||
AK087507.1 | - | 4 | 799 | |||
AK157206.1 | - | 4 | 848 | |||
AK170985.1 | - | 4 | 940 | |||
BC046510.1 | - | 4 | 897 | 4035182 | ||
BC094562.1 | - | 4 | 1354 | |||
U62559.1 | - | 3 | 453 |