Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009205.2 | NP_033231.2 | 2 | 611 | ||
GenBank mRNA | AK052623.1 | - | 2 | 582 | ||
AK132275.1 | - | 2 | 583 | |||
BC013441.1 | - | 2 | 553 | 4222972 | ||
CA493293.1 | - | 2 | 656 |