Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009205.2 | NP_033231.2 | 4 | 937 | ||
GenBank mRNA | AK052623.1 | - | 4 | 908 | ||
AK132275.1 | - | 4 | 909 | |||
BC013441.1 | - | 4 | 879 | 4222972 |