Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slit3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011412.3 | NP_035542.2 | 11 | 1083 | ||
GenBank mRNA | AF144629.1 | - | 11 | 1024 | ||
AK147292.1 | - | 11 | 1084 | |||
BC150780.1 | - | 11 | 1060 | 9087691 |