Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ILF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267809.1 | NP_001254738.1 | 8, 9 | 706 | ||
NM_004515.3 | NP_004506.2 | 8, 9 | 687 | |||
GenBank mRNA | AK223326.1 | - | 8, 9 | 607 | ||
AK302214.1 | - | 8, 9 | 650 | |||
AK313364.1 | - | 8, 9 | 646 | |||
BC000382.2 | - | 8, 9 | 612 | 2820505 | ||
BM800544.1 | - | 8, 9 | 662 | |||
HQ322261.1 | - | 7, 8 | 581 | |||
HQ322262.1 | - | 7, 8 | 581 | |||
U10323.1 | - | 8, 9 | 620 |