Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarca4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174078.1 | NP_001167549.1 | 15 | 2515 | ||
NM_001174079.1 | NP_001167550.1 | 15 | 2515 | |||
NM_011417.3 | NP_035547.2 | 15 | 2515 | |||
GenBank mRNA | AK141517.1 | - | 12 | 1669 | ||
AK147285.1 | - | 15 | 2469 | |||
AK160825.1 | - | 15 | 2419 | |||
AK166837.1 | - | 15 | 2420 | |||
BC060229.1 | - | 15 | 2409 | |||
BC079560.1 | - | 15 | 2515 | 30533489 | ||
S68108.1 | - | 5 | 481 |