Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011419.3 | NP_035549.1 | 7 | 1055 | ||
GenBank mRNA | AB221607.1 | - | 7 | 881 | ||
AF127244.1 | - | 7 | 893 | |||
AK047790.1 | - | 7 | 1056 | |||
AK140971.1 | - | 7 | 1050 | |||
AK141096.1 | - | 7 | 1051 | |||
BC060077.1 | - | 1034 | ||||
Z29652.1 | - | 7 | 811 |