Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252629.1 | NP_001239558.1 | 4 | 422 | ||
NM_011420.2 | NP_035550.1 | 4 | 358 | |||
GenBank mRNA | AK144778.1 | - | 4 | 359 | ||
AK152495.1 | - | 4 | 332 | |||
AK167832.1 | - | 4 | 356 | |||
BC045158.1 | - | 4 | 322 | 6486685 | ||
EU234021.1 | - | 4 | 422 | |||
U63294.1 | - | 4 | 336 | |||
U77714.1 | - | 4 | 319 | |||
Y12835.1 | - | 4 | 344 |