Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ILF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137673.1 | NP_001131145.1 | 12, 13 | 1698 | ||
NM_004516.3 | NP_004507.2 | 12, 13 | 1698 | |||
NM_012218.3 | NP_036350.2 | 12, 13 | 1698 | |||
NM_017620.2 | NP_060090.2 | 12, 13 | 1698 | |||
NM_153464.2 | NP_703194.1 | 12, 13 | 1698 | |||
XM_006722742.2 | XP_006722805.1 | 12, 13 | 1613 | |||
XM_011527984.2 | XP_011526286.2 | 12, 13 | 1731 | |||
XM_011527985.2 | XP_011526287.1 | 12, 13 | 1613 | |||
XM_011527987.1 | XP_011526289.1 | 8, 9 | 992 | |||
XM_017026763.1 | XP_016882252.1 | 13, 14 | 1822 | |||
XM_017026764.1 | XP_016882253.1 | 9, 10 | 1286 | |||
XM_017026765.1 | XP_016882254.1 | 11, 12 | 1515 | |||
GenBank mRNA | AB209062.1 | - | 11, 12 | 2318 | ||
AF141870.1 | - | 12, 13 | 1492 | |||
AF147209.1 | - | 10, 11 | 1393 | |||
AF167569.1 | - | 10, 11 | 1381 | |||
AF167570.1 | - | 10, 11 | 1381 | |||
AJ271744.1 | - | 8, 9 | 981 | |||
AJ271745.1 | - | 11, 12 | 1470 | |||
AJ271746.1 | - | 11, 12 | 1450 | |||
AK291205.1 | - | 12, 13 | 1597 | |||
AK291617.1 | - | 12, 13 | 1583 | |||
AK293170.1 | - | 4, 5 | 430 | |||
AK294084.1 | - | 8, 9 | 1001 | |||
BC064836.1 | - | 12, 13 | 1631 | 6503234 | ||
HQ322263.1 | - | 10, 11 | 1381 | |||
HQ322264.1 | - | 10, 11 | 1381 | |||
HQ322265.1 | - | 10, 11 | 1381 | |||
U10324.1 | - | 12, 13 | 1648 | |||
X98265.1 | - | 11, 12 | 1590 |