Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011421.2 | NP_035551.1 | 3 | 1308 | ||
GenBank mRNA | AK088147.1 | - | 3 | 1277 | ||
AK138133.1 | - | 2 | 1754 | |||
AK145534.1 | - | 3 | 1277 | |||
AK145702.1 | - | 3 | 1309 | |||
AK148189.1 | - | 3 | 1282 | |||
AK149579.1 | - | 3 | 1277 | |||
AK164167.1 | - | 3 | 1277 | |||
AK164717.1 | - | 3 | 1232 | |||
AK167802.1 | - | 3 | 1231 | |||
AK188573.1 | - | 1 | 17 | |||
AK191140.1 | - | 1 | 17 | |||
AK191382.1 | - | 1 | 17 | |||
AK192377.1 | - | 1 | 17 | |||
AK192380.1 | - | 1 | 17 | |||
AK195024.1 | - | 1 | 17 | |||
AK196690.1 | - | 1 | 17 | |||
AK198945.1 | - | 1 | 17 | |||
AK199356.1 | - | 1 | 17 | |||
AK199702.1 | - | 1 | 17 | |||
AK200055.1 | - | 1 | 17 | |||
AK200104.1 | - | 1 | 17 | |||
AK201352.1 | - | 1 | 17 | |||
AK202049.1 | - | 1 | 17 | |||
AK202792.1 | - | 17 | ||||
AK203823.1 | - | 1 | 17 | |||
AK205083.1 | - | 1 | 17 | |||
AK218218.1 | - | 1 | 17 | |||
BC011304.1 | - | 3 | 1243 | 3964394 | ||
BC034515.1 | - | 3 | 1233 | 4482098 | ||
Z14252.1 | - | 3 | 1254 |