Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sstr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009218.3 | NP_033244.2 | 3 | 638 | ||
GenBank mRNA | AK136904.1 | - | 3 | 639 | ||
BC120843.1 | - | 1 | 281 | 40129766 | ||
BC137670.1 | - | 1 | 281 | 8860689 |
Set Membership: |
G-protein coupled receptor |