Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sumo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301671.1 | NP_001288600.1 | 2 | 440 | ||
NM_001301672.1 | NP_001288601.1 | 2 | 314 | |||
NM_001301673.1 | NP_001288602.1 | 2 | 323 | |||
NM_019929.4 | NP_064313.1 | 2 | 314 | |||
GenBank mRNA | AF063847.1 | - | 2 | 141 | ||
AK012742.1 | - | 2 | 189 | |||
AK148306.1 | - | 2 | 178 | |||
AK150063.1 | - | 2 | 187 | |||
AK150200.1 | - | 2 | 187 | |||
AK160169.1 | - | 2 | 186 | |||
AK162035.1 | - | 2 | 187 | |||
AK167207.1 | - | 2 | 155 | |||
AK168988.1 | - | 2 | 178 | |||
AK170896.1 | - | 2 | 372 | |||
AK171169.1 | - | 2 | 372 | |||
AK186890.1 | - | 1 | 26 | |||
BC115488.1 | - | 2 | 121 | 40100722 | ||
BC115489.1 | - | 2 | 121 | 40100723 | ||
BY085566.1 | - | 2 | 188 | |||
CJ069985.1 | - | 2 | 216 | |||
CJ143013.1 | - | 2 | 417 | |||
CJ159246.1 | - | 2 | 231 | |||
CJ187500.1 | - | 3 | 407 | |||
CK619326.1 | - | 2 | 174 |