Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snap25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291056.1 | NP_001277985.1 | 4 | 352 | ||
NM_011428.3 | NP_035558.1 | 4 | 352 | |||
GenBank mRNA | AF483516.1 | - | 3 | 139 | ||
AF483517.1 | - | 3 | 139 | |||
AK078038.1 | - | 4 | 350 | |||
AU080862.1 | - | 4 | 319 | |||
BC018249.1 | - | 4 | 240 | 4504644 | ||
M22012.1 | - | 4 | 302 |