Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snap25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011428.3 | NP_035558.1 | 5 | 408 | ||
GenBank mRNA | AF483516.1 | - | 4 | 195 | ||
AF483517.1 | - | 4 | 195 | |||
AK078038.1 | - | 5 | 406 | |||
BC018249.1 | - | 5 | 296 | 4504644 | ||
M22012.1 | - | 5 | 358 |