Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005045.2 | - | 7 | 646 | ||
AK011564.1 | - | 7 | 602 | |||
AK012272.1 | - | 5 | 295 | |||
AK014311.2 | - | 7 | 643 | |||
AK077425.1 | - | 7 | 648 | |||
AK133115.1 | - | 7 | 647 | |||
AK146729.1 | - | 7 | 647 | |||
AK148013.1 | - | 7 | 673 | |||
BC043938.1 | - | 1 | 448 | |||
BC049128.1 | - | 7 | 612 | |||
BC051414.1 | - | 6 | 447 | |||
BC131994.1 | - | 7 | 539 | 40130271 | ||
BC132020.1 | - | 7 | 539 | 40130297 |