Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009226.4 | NP_033252.1 | 3 | 317 | ||
GenBank mRNA | AK005517.1 | - | 3 | 307 | ||
AK007674.1 | - | 3 | 318 | |||
AK011455.1 | - | 3 | 311 | |||
AK013075.1 | - | 3 | 284 | |||
AK134582.1 | - | 3 | 311 | |||
AK135979.1 | - | 3 | 307 | |||
BC024875.1 | - | 3 | 303 | 4009014 | ||
M58558.1 | - | 3 | 246 |