Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrpe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009227.3 | NP_033253.1 | 3 | 197 | ||
GenBank mRNA | AK019462.1 | - | 3 | 181 | ||
BC008262.1 | - | 3 | 173 | 2650413 | ||
BC051207.1 | - | 3 | 155 | 1396204 | ||
BC055765.1 | - | 3 | 139 | 6407944 | ||
BY168551.1 | - | 3 | 197 | |||
X65704.1 | - | 3 | 101 |