Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sntb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009229.4 | NP_033255.1 | 3 | 968 | ||
GenBank mRNA | AK040423.1 | - | 3 | 969 | ||
AK080059.1 | - | 3 | 965 | |||
AK131687.1 | - | 3 | 993 | |||
AK165009.1 | - | 3 | 970 | |||
AK165900.1 | - | 3 | 945 | |||
BC138850.1 | - | 3 | 967 | 8861872 | ||
BC145442.1 | - | 3 | 967 | 9054000 | ||
U00678.1 | - | 3 | 967 |