Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011435.3 | NP_035565.1 | 2 | 370 | ||
GenBank mRNA | AF076286.1 | - | 1 | 132 | ||
AK075570.1 | - | 2 | 225 | |||
AK145194.1 | - | 229 | ||||
BC010975.1 | - | 2 | 188 | 3983917 | ||
D50856.1 | - | 1 | 123 | |||
U38261.1 | - | 2 | 372 |