Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011435.3 | NP_035565.1 | 2 | 397 | ||
GenBank mRNA | AF076286.1 | - | 1 | 159 | ||
AK075570.1 | - | 2 | 252 | |||
AK145194.1 | - | 256 | ||||
BC010975.1 | - | 2 | 215 | 3983917 | ||
D50856.1 | - | 1 | 150 | |||
U38261.1 | - | 2 | 399 |