Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sorl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011436.3 | NP_035566.2 | 42 | 5833 | ||
GenBank mRNA | AB015790.1 | - | 42 | 5657 | ||
AF031816.1 | - | 5111 | ||||
AK088154.1 | - | 421 | ||||
AK147303.1 | - | 42 | 5834 | |||
AK169831.1 | - | 421 |