Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sos2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135559.1 | NP_001129031.1 | 10 | 1965 | ||
GenBank mRNA | Z11664.1 | - | 9 | 1609 |