Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPDH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000883.3 | NP_000874.2 | 8 | 829 | ||
NM_001102605.1 | NP_001096075.1 | 7 | 799 | |||
NM_001142573.1 | NP_001136045.1 | 5 | 599 | |||
NM_001142574.1 | NP_001136046.1 | 5 | 584 | |||
NM_001142575.1 | NP_001136047.1 | 4 | 524 | |||
NM_001142576.1 | NP_001136048.1 | 7 | 730 | |||
NM_001304521.1 | NP_001291450.1 | 5 | 627 | |||
NM_183243.2 | NP_899066.1 | 6 | 721 | |||
XM_017012168.1 | XP_016867657.1 | 7 | 799 | |||
XM_017012169.1 | XP_016867658.1 | 6 | 721 | |||
XM_017012170.1 | XP_016867659.1 | 5 | 628 | |||
XM_017012171.1 | XP_016867660.1 | 7 | 917 | |||
XM_017012172.1 | XP_016867661.1 | 7 | 917 | |||
XM_017012173.1 | XP_016867662.1 | 7 | 799 | |||
GenBank mRNA | AK054640.1 | - | 4 | 496 | ||
AK054667.1 | - | 6 | 721 | |||
AK092452.1 | - | 7 | 766 | |||
AK122994.1 | - | 7 | 798 | |||
AK131294.1 | - | 5 | 546 | |||
AK293413.1 | - | 5 | 556 | |||
AK297943.1 | - | 5 | 571 | |||
BC033622.1 | - | 6 | 712 | 5210847 | ||
BX406207.2 | - | 6 | 724 | |||
CD014008.1 | - | 6 | 513 | |||
CD014010.1 | - | 7 | 613 | |||
CR933672.1 | - | 765 | ||||
J05272.1 | - | 8 | 1093 |