Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009238.2 | NP_033264.2 | 1 | 1814 | ||
GenBank mRNA | AK028989.1 | - | 1830 | |||
AK053840.1 | - | 1 | 1832 | |||
BC052736.1 | - | 1 | 1814 | 6822618 | ||
BC060669.1 | - | 1 | 1935 | |||
X70298.1 | - | 1 | 1809 |