Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079875.2 | NP_001073344.1 | 5 | 1713 | ||
NM_009241.3 | NP_033267.2 | 4 | 1707 | |||
GenBank mRNA | AK005638.1 | - | 5 | 1700 | ||
AY228460.1 | - | 3 | 1388 | |||
AY920282.1 | - | 4 | 1707 | |||
AY920283.1 | - | 5 | 1686 | |||
U33958.1 | - | 4 | 1697 |