Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sphk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172472.1 | NP_001165943.1 | 6 | 1373 | ||
NM_001172473.1 | NP_001165944.1 | 6 | 1259 | |||
NM_001172475.1 | NP_001165946.1 | 6 | 1298 | |||
NM_011451.3 | NP_035581.1 | 4 | 1030 | |||
NM_025367.6 | NP_079643.2 | 6 | 1256 | |||
GenBank mRNA | AF068748.1 | - | 6 | 1298 | ||
AF068749.1 | - | 4 | 1030 | |||
AF415213.1 | - | 4 | 928 | |||
AK028667.1 | - | 3 | 3564 | |||
AK155332.1 | - | 6 | 1358 | |||
AK160900.1 | - | 6 | 1249 | |||
BC011432.1 | - | 1 | 1630 | |||
BC037710.1 | - | 6 | 1171 | 4221357 |