Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011454.1 | NP_035584.1 | 5 | 503 | ||
GenBank mRNA | AK078753.1 | - | 5 | 573 | ||
AK154048.1 | - | 5 | 517 | |||
AK171741.1 | - | 4 | 340 | |||
BC061050.1 | - | 5 | 510 | 30276988 | ||
U96707.2 | - | 5 | 503 |