Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina3n | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009252.2 | NP_033278.2 | 5 | 1294 | ||
GenBank mRNA | BC013651.1 | - | 5 | 1273 | 4160649 | |
M64086.1 | - | 5 | 1249 |