Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002164.5 | NP_002155.1 | 2 | 254 | ||
GenBank mRNA | AK313259.1 | - | 2 | 235 | ||
AU137351.1 | - | 2 | 254 | |||
AY221100.1 | - | 2 | 140 | |||
BC027882.1 | - | 2 | 185 | 5208340 | ||
CD690354.1 | - | 2 | 259 | |||
JF772862.1 | - | 2 | 140 | |||
M34455.1 | - | 2 | 779 | |||
X17668.1 | - | 2 | 162 |