Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011459.4 | NP_035589.1 | 7 | 1038 | ||
GenBank mRNA | AK032659.1 | - | 7 | 1037 | ||
AK036811.1 | - | 7 | 974 | |||
AK148413.1 | - | 7 | 1036 | |||
BC011076.1 | - | 7 | 1018 | |||
BC025121.1 | - | 7 | 1028 | 4487039 | ||
U96703.2 | - | 7 | 1029 |
Gene Symbol: Spi16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U96702.1 | - | 1 | 57 |