Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283028.1 | NP_001269957.1 | 5 | 883 | ||
NM_001283029.1 | NP_001269958.1 | 5 | 748 | |||
NM_001283030.1 | NP_001269959.1 | 4 | 607 | |||
NM_011462.3 | NP_035592.1 | 3 | 512 | |||
NM_146043.4 | NP_666155.1 | 5 | 808 | |||
GenBank mRNA | AK088127.1 | - | 5 | 749 | ||
AK134259.1 | - | 5 | 771 | |||
AK135816.1 | - | 3 | 507 | |||
AK135847.1 | - | 3 | 511 | |||
AK135933.1 | - | 3 | 510 | |||
AK136233.1 | - | 3 | 510 | |||
AK136251.1 | - | 3 | 511 | |||
AK139899.1 | - | 3 | 509 | |||
AK162171.1 | - | 3 | 509 | |||
AK162195.1 | - | 3 | 509 | |||
AK162258.1 | - | 3 | 506 | |||
AK163263.1 | - | 3 | 508 | |||
AK163293.1 | - | 3 | 509 | |||
AK163301.1 | - | 3 | 513 | |||
AK163308.1 | - | 3 | 511 | |||
AK163382.1 | - | 3 | 509 | |||
AK166528.1 | - | 3 | 512 | |||
AK166532.1 | - | 3 | 512 | |||
AK166580.1 | - | 3 | 471 | |||
AK166677.1 | - | 3 | 508 | |||
AK168544.1 | - | 5 | 809 | |||
BC016517.1 | - | 5 | 794 | 2654233 | ||
U48972.1 | - | 3 | 506 |