Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spink4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011463.2 | NP_035593.2 | 3, 4 | 253 | ||
GenBank mRNA | BC117048.1 | - | 3, 4 | 256 | 40126367 | |
BC117050.1 | - | 3, 4 | 255 | 40126369 | ||
CF381698.1 | - | 3, 4 | 259 | |||
Y11505.1 | - | 317 |